Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex 16-Turn Valve Cartridge, Range 3 for Al Frame Systems
Catalog No. 501961093
Encompass
Select a valve-cartridge that matches your frame. Insert these needle valves into the flowmeter frames to control flow rate. Choose a 16-turn, high-resolution metering valve-cartridge for greater precision of flow rate. If your flowtube is in units other than mL/min, convert them to mL/min and choose a valve within that flow range from the table below. See website and virtual catalog page for more details.
Catalog No. 50-196-1093 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0321861
Select a valve-cartridge that matches your frame. Insert these needle valves into the flowmeter frames to control flow rate. Choose a 16-turn, high-resolution metering valve-cartridge for greater precision of flow rate. If your flowtube is in units other than mL/min, convert them to mL/min and choose a valve within that flow range from the table below. See website and virtual catalog page for more details.