Learn More
Waters Corp Cartridge Vent Valve, bioQSM, ACQUITY, UPLC
Catalog No. 502390867 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This is a replacement Vent Valve Cartridge designed for use in specific solvent managers. It is compatible with the H-Class Bio Quaternary Solvent Manager and both Binary and Auxiliary Solvent Managers in the M-Class series.
  • Replacement Vent Valve Cartridge.
  • Designed for use in specific solvent managers.
  • Compatible with H-Class Bio Quaternary Solvent Manager and M-Class Binary and Auxiliary Solvent Managers.
  • System type: UPLC.
  • Units per package: 1.
  • Replacement schedule: As required.
  • Functions as a component within BSM and ISM systems.
Catalog No. 50-239-0867 Supplier Waters Corp Supplier No. 700005413
This is a replacement Vent Valve Cartridge designed for use in specific solvent managers. It is compatible with the H-Class Bio Quaternary Solvent Manager and both Binary and Auxiliary Solvent Managers in the M-Class series.
  • Replacement Vent Valve Cartridge.
  • Designed for use in specific solvent managers.
  • Compatible with H-Class Bio Quaternary Solvent Manager and M-Class Binary and Auxiliary Solvent Managers.
  • System type: UPLC.
  • Units per package: 1.
  • Replacement schedule: As required.
  • Functions as a component within BSM and ISM systems.