Learn More
Grainger Honeywell North Cartridge, Acid Gas, Threaded, White, 1 Pair
Catalog No. 502790787
Encompass_Preferred
This acid gas respirator cartridge provides reliable respiratory protection against various hazardous acid gases and formaldehyde. Designed for use in industrial and laboratory settings, it ensures safety in environments where exposure to corrosive chemical vapors is a concern.
  • NIOSH approved for protection against chlorine, hydrogen chloride, sulfur dioxide, hydrogen sulfide, hydrogen fluoride, chlorine dioxide, and formaldehyde.
  • Features a durable, lightweight design with a threaded connection for secure attachment to compatible air-purifying respirators.
  • White color-coded for easy identification in compliance with respiratory protection standards.
  • Easy to install and replace in the field.
  • Suitable for a wide range of applications including chemical processing, metal treatment, and industrial maintenance.
Catalog No. 50-279-0787 Supplier Grainger Supplier No. 16M238
This acid gas respirator cartridge provides reliable respiratory protection against various hazardous acid gases and formaldehyde. Designed for use in industrial and laboratory settings, it ensures safety in environments where exposure to corrosive chemical vapors is a concern.
  • NIOSH approved for protection against chlorine, hydrogen chloride, sulfur dioxide, hydrogen sulfide, hydrogen fluoride, chlorine dioxide, and formaldehyde.
  • Features a durable, lightweight design with a threaded connection for secure attachment to compatible air-purifying respirators.
  • White color-coded for easy identification in compliance with respiratory protection standards.
  • Easy to install and replace in the field.
  • Suitable for a wide range of applications including chemical processing, metal treatment, and industrial maintenance.