Learn More
3M™ 6000 Series Cartridges
NIOSH-approved cartridges for use with 3M™ full and half facepiece respirators 6000, 7000 and FF-400 Series.
Supplier:  3M™ 7000051850
 View more versions of this product
Catalog No. 17-986-11B
Add to Cart
This item is not returnable. View return policy
Description
Description
NIOSH-approved cartridges for use with 3M™ full and half facepiece respirators 6000, 7000 and FF-400 Series.
  • For use with 3M full and half facepiece respirators 6000, 7000 and FF-400 Series
  • Bayonet cartridge connection
  • NIOSH-approved for protection against a variety of gases, vapors, and particulate hazards*
  • For contaminants up to 10X the Permissible Exposure Limit (PEL) with half-facepiece respirators or 50X the PEL (when fit tested) with full-facepiece respirators or according to specific OSHA standards or applicable government regulations, whichever is lower
  • Not intended for use in atmospheres that are unknown or immediately dangerous to life or death (IDLH)
  • Do not exceed maximum use concentrations established by regulatory standards
  • These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product
  • Misuse may result in sickness or death
*Details found at individual product level according
Specifications
Specifications
Acid Gases: Chlorine, Hydrogen Chrloride, Sulfur Dioxide, Chlorine Dioxide, Hydrogen Sulfide
Bayonet
Cartridge
3M™ half and full facepiece 6000, 7000, ff-400 series respirators
60/Cs.
10.6 cm
8.6 cm
NIOSH-approved
White
Chemical manufacturing, glass etching, pulp and paper and laboratories
6002
4.2 in.
3.4 in.
Product Suggestions
Product Suggestions