Learn More
Teledyne ISCO, Inc Empty solid load cartridges. Sample load, empty Rf cartridges (holds up to 65 gram) pkg. of 12.
Catalog No. 50871518 Shop All Teledyne ISCO Inc. Products
Encompass
Solid Load Cartridges. Available prepacked or empty to fill with your own sorbent. Solid sample loading is a useful technique to load samples to be purified with a CombiFlash instrument onto a RediSep Rf column, particularly in the case of low-solubility samples. All CombiFlash systems are designed for either solid or liquid sample loading. Teledyne Isco Solid Phase Sample Load Cartridges are available either empty (to fill with your own sorbent) or pre-filled with silica gel or reversed-phase C18 material. Several sizes of sample load cartridges (5, 25, 65, and 270 gram nominal sorbent capacity) allow loading of sample amounts from milligrams to tens of grams. When used on the CombiFlash Companion and Rf systems, the cartridges are used with a solid load cartridge cap. This adjustable cap minimizes void volumes in the cartridge so that accurate gradients can be delivered.
Catalog No. 50-871-518 Supplier Teledyne ISCO, Inc Supplier No. 693873225
Solid Load Cartridges. Available prepacked or empty to fill with your own sorbent. Solid sample loading is a useful technique to load samples to be purified with a CombiFlash instrument onto a RediSep Rf column, particularly in the case of low-solubility samples. All CombiFlash systems are designed for either solid or liquid sample loading. Teledyne Isco Solid Phase Sample Load Cartridges are available either empty (to fill with your own sorbent) or pre-filled with silica gel or reversed-phase C18 material. Several sizes of sample load cartridges (5, 25, 65, and 270 gram nominal sorbent capacity) allow loading of sample amounts from milligrams to tens of grams. When used on the CombiFlash Companion and Rf systems, the cartridges are used with a solid load cartridge cap. This adjustable cap minimizes void volumes in the cartridge so that accurate gradients can be delivered.