Learn More
CELLINK LLC CARTRIDGES,EMPTY,W/O END, 3ML
SDP
Catalog No. NC2143455
Encompass
Empty cartridges without end and tip caps, 3 mLEmpty, autoclavable cartridges with pistons for bioprinting.
This item is not returnable. View return policy
Catalog No. NC2143455 Supplier CELLINK LLC Supplier No. CSC010300502
Empty cartridges without end and tip caps, 3 mLEmpty, autoclavable cartridges with pistons for bioprinting.