Learn More
Eldex Laboratories Inc High Temperature Burner for Cartridge
Catalog No. NC0513474
Encompass_Preferred
Piezo electric ignition, needle valve, and T=1600 degrees celsius.
Catalog No. NC0513474 Supplier Eldex Laboratories Inc Supplier No. 1424
Piezo electric ignition, needle valve, and T=1600 degrees celsius.