Learn More
Bio Rad Laboratories CARTRIDGES MICRO-GUARD ANION
Catalog No. 509012176
Encompass
Cartridges, Anion CO3 -; Bio-Rad; For Aminex silver and lead form columns; 30 x 4.6mm; Quantity: 2
Catalog No. 50-901-2176 Supplier Bio Rad Laboratories Supplier No. 1250506
Cartridges, Anion CO3 -; Bio-Rad; For Aminex silver and lead form columns; 30 x 4.6mm; Quantity: 2