Learn More
Bio Rad Laboratories CARTRIDGES REFILL MICRO-GUA
Catalog No. 509010096
Encompass
Cartridge, Refill; Bio-Rad; Micro-Guard; Cation H; For Aminex hydrogen-form columns; Ionic form: Hydrogen; 30 x 4.6mm, 2; pH range 1-3
Catalog No. 50-901-0096 Supplier Bio Rad Laboratories Supplier No. 1250129
Cartridge, Refill; Bio-Rad; Micro-Guard; Cation H; For Aminex hydrogen-form columns; Ionic form: Hydrogen; 30 x 4.6mm, 2; pH range 1-3