Learn More
Bio Rad Laboratories CARTRIDGES REFILL MICRO-GUA
Supplier:  Bio Rad Laboratories 1250119
Encompass
Cartridges, Carbo-P Refill; Bio-Rad; For Aminex HPX-87P column; 30 x 4.6mm; pH range 6-8; Quantity: 2
Catalog No. 50-901-2173