Learn More
Bio Rad Laboratories CARTRIDGES REFILL MICRO-GUA
Supplier:  Bio Rad Laboratories 1250118
Encompass
Cartridges, De-Ashing Refill; Bio-Rad; Micro-Guard; For Aminex silver and lead form columns; 30 x 4.6mm; pH range 6-8; Quantity: 2
Catalog No. 50-901-2172