Learn More
Teledyne ISCO, Inc RediSep prepacked silica gel disposable sample load cartridges, 25 gram.  Package of 15
Catalog No. 509018246 Shop All Teledyne ISCO Inc. Products
Encompass
Polypropylene cartridges available either prepacked with normal-phase or reversed-phase sorbent, or empty, to be filled with user-supplied sorbent.
Catalog No. 50-901-8246 Supplier Teledyne ISCO, Inc Supplier No. 693873241
Polypropylene cartridges available either prepacked with normal-phase or reversed-phase sorbent, or empty, to be filled with user-supplied sorbent.