Learn More
Medchemexpress LLC Carubicin hydrochloride | 52794-97-5 | 98.3% | 5 MG
SDP
Catalog No. 5000409649
Encompass_Preferred
Carubicin hydrochloride is a microbially-derived compound with potential for cancer disease research. It is an effective inhibitor of VHL-defective (VHL-/-) CCRCC cell proliferation and induces apoptosis through a mechanism independent of p53 or hypoxia-inducible factor HIF2.
  • Effective inhibitor of VHL-defective (VHL-/-) CCRCC cell proliferation
  • Induces apoptosis independent of p53 or hypoxia-inducible factor HIF2
  • Suitable for research of cancer diseases
Catalog No. 50-004-09649 Supplier Medchemexpress LLC Supplier No. HYB2171A5MG
Carubicin hydrochloride is a microbially-derived compound with potential for cancer disease research. It is an effective inhibitor of VHL-defective (VHL-/-) CCRCC cell proliferation and induces apoptosis through a mechanism independent of p53 or hypoxia-inducible factor HIF2.
  • Effective inhibitor of VHL-defective (VHL-/-) CCRCC cell proliferation
  • Induces apoptosis independent of p53 or hypoxia-inducible factor HIF2
  • Suitable for research of cancer diseases