Learn More
Medchemexpress LLC Carubicin hydrochloride | 52794-97-5 | 98.3% | 50 MG
SDP
Catalog No. 5000409978
Encompass_Preferred
Carubicin hydrochloride is a microbially-derived compound with potential for research related to cancer diseases.
  • Inhibits VHL-defective CCRCC cell proliferation.
  • Induces apoptosis independent of p53 or HIF2.
  • Derived from microorganisms (bacterium Actinomadura carminata).
Catalog No. 50-004-09978 Supplier Medchemexpress LLC Supplier No. HYB2171A50MG
Carubicin hydrochloride is a microbially-derived compound with potential for research related to cancer diseases.
  • Inhibits VHL-defective CCRCC cell proliferation.
  • Induces apoptosis independent of p53 or HIF2.
  • Derived from microorganisms (bacterium Actinomadura carminata).