Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Carubicin (hydrochloride) | 52794-97-5 | 98.3% | 549.95 | C26H28ClNO10 | 10 MG
SDP

Catalog No. 5000298880
Encompass_Preferred

Carubicin (hydrochloride) is the hydrochloride salt of a microbially derived anthracycline supplied for research use. It has been reported to inhibit proliferation of VHL-defective clear cell renal cell carcinoma cells and to induce apoptosis by a mechanism independent of p53 and HIF2.

  • Chemical formula C26H28ClNO10.
  • Cas number 52794-97-5.
  • Molecular weight 549.95 g/mol.
  • Purity 98.33% for research applications.
  • Supplied as a solid in milligram quantities suitable for in vitro assays.

Catalog No. 50-002-98880 Supplier Medchemexpress LLC Supplier No. HYB2171A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Carubicin (hydrochloride) is the hydrochloride salt of a microbially derived anthracycline supplied for research use. It has been reported to inhibit proliferation of VHL-defective clear cell renal cell carcinoma cells and to induce apoptosis by a mechanism independent of p53 and HIF2.

  • Chemical formula C26H28ClNO10.
  • Cas number 52794-97-5.
  • Molecular weight 549.95 g/mol.
  • Purity 98.33% for research applications.
  • Supplied as a solid in milligram quantities suitable for in vitro assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.