Learn More
Medchemexpress LLC HY-N0711 100mg Medchemexpress, Carvacrol CAS:499-75-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN0711100MG
Encompass_Preferred
Medchemexpress, HY-N0711 100mg Carvacrol CAS:499-75-2 Carvacrol is a monoterpenoid phenol isolated from Lamiaceae family plants, with antioxidant, anti-inflammatory and anticancer properties. Carvacrol causes cell cycle arrest in G0/G1, downregulates Notch-1, and Jagged-1, and induces apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0306