Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Carvacrol methyl ether | 6379-73-3 | 99.9% | 164.25 | C11H16O | 1 G
SDP

Catalog No. 5000209800
Encompass_Preferred

Carvacrol methyl ether is the methyl ether derivative of carvacrol, isolated from plant volatile oils and used in research for its antibacterial properties and proton-acceptor behavior.

  • Exhibits antibacterial activity.
  • Acts as a proton acceptor.
  • Liquid form, colorless to light yellow in appearance.
  • High purity (approximately 99.9%).
  • Molecular formula C11H16O; mol wt 164.25.
  • Storage: -20°C for up to 3 years or 4°C for 2 years for pure material; -80°C for 6 months or -20°C for 1 month in solvent.
  • Available in multiple pack sizes including 1 g.

Catalog No. 50-002-09800 Supplier Medchemexpress LLC Supplier No. HYW0499701G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Carvacrol methyl ether is the methyl ether derivative of carvacrol, isolated from plant volatile oils and used in research for its antibacterial properties and proton-acceptor behavior.

  • Exhibits antibacterial activity.
  • Acts as a proton acceptor.
  • Liquid form, colorless to light yellow in appearance.
  • High purity (approximately 99.9%).
  • Molecular formula C11H16O; mol wt 164.25.
  • Storage: -20°C for up to 3 years or 4°C for 2 years for pure material; -80°C for 6 months or -20°C for 1 month in solvent.
  • Available in multiple pack sizes including 1 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.