Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Carvacrol methyl ether | 6379-73-3 | 99.9% | 164.25 | C11H16O | 1 ML
SDP

Catalog No. 5000210536
Encompass_Preferred

Carvacrol methyl ether is a methyl ether derivative of carvacrol supplied as a research-grade reagent and analytical standard. It exhibits antibacterial activity and is available as a 10 mM solution in DMSO (1 mL) or as solids in multiple sizes for laboratory research purposes.

  • Antibacterial activity observed in vitro.
  • Available as 10 mM solution in DMSO (1 mL) and as solids (100 mg, 250 mg, 1 g, larger sizes on request).
  • High purity (approximately 99.9%).
  • Molecular formula C11H16O; molecular weight 164.25.
  • Recommended storage: solids at -20°C (long term); solutions at -80°C for extended storage.

Catalog No. 50-002-10536 Supplier Medchemexpress LLC Supplier No. HYW04997010MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Carvacrol methyl ether is a methyl ether derivative of carvacrol supplied as a research-grade reagent and analytical standard. It exhibits antibacterial activity and is available as a 10 mM solution in DMSO (1 mL) or as solids in multiple sizes for laboratory research purposes.

  • Antibacterial activity observed in vitro.
  • Available as 10 mM solution in DMSO (1 mL) and as solids (100 mg, 250 mg, 1 g, larger sizes on request).
  • High purity (approximately 99.9%).
  • Molecular formula C11H16O; molecular weight 164.25.
  • Recommended storage: solids at -20°C (long term); solutions at -80°C for extended storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.