Learn More
Medchemexpress LLC Carvacrol methyl ether | 6379-73-3 | MFCD00216689 | 99.9% | 164.25 g/mol | C11H16O | 250 MG
SDP
Catalog No. 5000209190
Encompass_Preferred
Carvacrol methyl ether is a naturally derived carvacrol analogue isolated from plant volatile oils. It exhibits antibacterial activity and acts as a proton acceptor; it is provided as a research-grade reagent in multiple formats for laboratory use.
  • Natural product derivative with antibacterial activity.
  • High purity suitable for analytical and biological research.
  • Available in solution and solid formats (for example, 10 mM in DMSO, 100 mg-10 g) for experimental flexibility.
  • Molecular weight 164.25 g/mol and formula C11H16O for easy reference in calculations.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-09190 Supplier Medchemexpress LLC Supplier No. 250MGHYW049970
Carvacrol methyl ether is a naturally derived carvacrol analogue isolated from plant volatile oils. It exhibits antibacterial activity and acts as a proton acceptor; it is provided as a research-grade reagent in multiple formats for laboratory use.
  • Natural product derivative with antibacterial activity.
  • High purity suitable for analytical and biological research.
  • Available in solution and solid formats (for example, 10 mM in DMSO, 100 mg-10 g) for experimental flexibility.
  • Molecular weight 164.25 g/mol and formula C11H16O for easy reference in calculations.
  • Intended for research use only; not for human or clinical use.