Learn More
Indofine Chemical Carvacrol With Gc, 1 Gm
SDP
Catalog No. 50908906
Encompass_Preferred
CARVACROL with GC 1 GM, Chromatography Standards, 2-Hydroxy-4-cymene; Isothymol, C10H14O, MW: 150.22, 98% by GC, 499-75-2, MFCD00002236
Catalog No. 50-908-906 Supplier Indofine Chemical Supplier No. 025440S 1 GM
CARVACROL with GC 1 GM, Chromatography Standards, 2-Hydroxy-4-cymene; Isothymol, C10H14O, MW: 150.22, 98% by GC, 499-75-2, MFCD00002236