Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aobchem Carvacryl acetate | ≥97% | CAS 6380-28-5 | C12H16O2 | 1g

Catalog No. 5000863192
Encompass_Preferred

Carvacryl acetate | ≥97% | CAS 6380-28-5 | C12H16O2 | 1g

Catalog No. 50-008-63192 Supplier Aobchem Supplier No. A3017571G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Carvacryl acetate | ≥97% | CAS 6380-28-5 | C12H16O2 | 1g

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.