Learn More
Apexbio Technology LLC Carvedilol 72956-09-3 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062306
Encompass_Preferred
Carvedilol is an antagonist targeting 1- and -adrenergic receptors employed in research on cardiovascular diseases especially congestive heart failure and hypertension Mechanistically carvedilol blocks adrenergic receptor-mediated signaling influencing sympathetic nervous system activity Additionally carvedilol exhibits antioxidative properties in vitro studies demonstrate suppression of Fe2 -induced lipid peroxidation (IC50 8 1 M) protection of -tocopherol depletion due to Fe2 exposure (IC50 17 6 M) and reduction of hydroxyl radical-derived DMPO-OH signals (IC50 25 M) It also inhibits proliferation (IC50 0 3-2 0 M) and migration (IC50 3 M) in vascular smooth muscle cells induced by platelet-derived growth factor supporting investigation of vascular remodeling processes
Catalog No. 50-000-62306 Supplier Apexbio Technology LLC Supplier No. B13325.1
Carvedilol is an antagonist targeting 1- and -adrenergic receptors employed in research on cardiovascular diseases especially congestive heart failure and hypertension Mechanistically carvedilol blocks adrenergic receptor-mediated signaling influencing sympathetic nervous system activity Additionally carvedilol exhibits antioxidative properties in vitro studies demonstrate suppression of Fe2 -induced lipid peroxidation (IC50 8 1 M) protection of -tocopherol depletion due to Fe2 exposure (IC50 17 6 M) and reduction of hydroxyl radical-derived DMPO-OH signals (IC50 25 M) It also inhibits proliferation (IC50 0 3-2 0 M) and migration (IC50 3 M) in vascular smooth muscle cells induced by platelet-derived growth factor supporting investigation of vascular remodeling processes