Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Carvedilol

Catalog No. AC462450050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC462450050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC462450050 Supplier Thermo Scientific Chemicals Supplier No. 462450050

missing translation for 'validMainframeMsg'

CAS: 72956-09-3 | C24H26N2O4 | 406.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72956-09-3
Molecular Formula C24H26N2O4
Molecular Weight (g/mol) 406.48
InChI Key OGHNVEJMJSYVRP-UHFFFAOYNA-N
IUPAC Name 1-(9H-carbazol-4-yloxy)-3-{[2-(2-methoxyphenoxy)ethyl]amino}propan-2-ol
SMILES COC1=CC=CC=C1OCCNCC(O)COC1=CC=CC2=C1C1=C(N2)C=CC=C1

Specifications

Melting Point 114.0°C to 115.0°C
Quantity 5 g
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: very soluble in dimethyl sulfoxide, soluble in methanol & methylene chloride, slightly soluble in ether, ethanol & isopropanol
Formula Weight 406.47
Chemical Name or Material Carvedilol
Hazard Statement GHS H Statement Toxic to aquatic life with long lasting effects.

Precautionary Statement GHS P Statement Avoid release to the environment.

Collect spillage.

Dispose of contents/container to an approved waste disposal plant.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.