Learn More
Medchemexpress LLC Carvedilol | 72956-09-3 | 99.98% | 1 ML
SDP
Catalog No. 5000432510
Encompass_Preferred
Carvedilol is a non-selective β/α-1 blocker. It inhibits lipid peroxidation in a dose-dependent manner with an IC50 of 5 μM. This multiple-action antihypertensive agent has potential use in angina and congestive heart failure. It also acts as an autophagy inducer that inhibits the NLRP3 inflammasome.
  • Non-selective β/α-1 blocker.
  • Inhibits lipid peroxidation with an IC50 of 5 μM.
  • Antihypertensive agent for angina and congestive heart failure.
  • Autophagy inducer.
  • Inhibits the NLRP3 inflammasome.
  • Inhibits superoxide generation by activated human neutrophils (IC50 of 28 μM).
  • Scavenges oxygen free radicals in a cell-free system (IC50 of 25 μM).
Catalog No. 50-004-32510 Supplier Medchemexpress LLC Supplier No. HYB000610MM/1ML
Add to Cart
Add to Cart
Carvedilol is a non-selective β/α-1 blocker. It inhibits lipid peroxidation in a dose-dependent manner with an IC50 of 5 μM. This multiple-action antihypertensive agent has potential use in angina and congestive heart failure. It also acts as an autophagy inducer that inhibits the NLRP3 inflammasome.
  • Non-selective β/α-1 blocker.
  • Inhibits lipid peroxidation with an IC50 of 5 μM.
  • Antihypertensive agent for angina and congestive heart failure.
  • Autophagy inducer.
  • Inhibits the NLRP3 inflammasome.
  • Inhibits superoxide generation by activated human neutrophils (IC50 of 28 μM).
  • Scavenges oxygen free radicals in a cell-free system (IC50 of 25 μM).