Learn More
American Radiolabeled Chemicals Inc CARVEDILOL ME-3H 50UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ART 178750UC
Encompass_Preferred
Carvedilol-methyl-3H 50uCi in ethanol, 1 mCi/ml, S.A. 80-85 Ci/mmol, M.W. 406.47, Shipped in Dry Ice, Exclusive
Catalog No. 50-901-215