Learn More
Apexbio Technology LLC Carvedilol Phosphate 610309-89-2 25g
SDP
Catalog No. 5000092696
Encompass_Preferred
Carvedilol phosphate (CAS 610309-89-2) is a non-selective beta-adrenoceptor and alpha-1 receptor antagonist targeting adrenergic receptors It is designed to inhibit sympathetic nervous system signaling thereby regulating cardiovascular function and suppressing lipid peroxidation Carvedilol phosphate exerts its biological activity primarily through blockade of beta and alpha-1 adrenergic receptors and inhibition of lipid peroxidation In in vitro studies carvedilol phosphate demonstrates autophagy induction and inhibition of NLRP3 inflammasome activation with reported IC50 values for beta-adrenergic blockade typically in the nanomolar-to-low micromolar range depending on receptor subtype and assay conditions Based on these pharmacological properties carvedilol phosphate holds research potential in hypertension congestive heart failure myocardial ischemia and inflammatory response studies
Catalog No. 50-000-92696 Supplier Apexbio Technology LLC Supplier No. C640425000
Carvedilol phosphate (CAS 610309-89-2) is a non-selective beta-adrenoceptor and alpha-1 receptor antagonist targeting adrenergic receptors It is designed to inhibit sympathetic nervous system signaling thereby regulating cardiovascular function and suppressing lipid peroxidation Carvedilol phosphate exerts its biological activity primarily through blockade of beta and alpha-1 adrenergic receptors and inhibition of lipid peroxidation In in vitro studies carvedilol phosphate demonstrates autophagy induction and inhibition of NLRP3 inflammasome activation with reported IC50 values for beta-adrenergic blockade typically in the nanomolar-to-low micromolar range depending on receptor subtype and assay conditions Based on these pharmacological properties carvedilol phosphate holds research potential in hypertension congestive heart failure myocardial ischemia and inflammatory response studies