Learn More
Medchemexpress LLC Carvedilol phosphate hemihydrate | 610309-89-2 | 99.9% | C24H26N2O4.H3PO4.1/2H2O | 1 G
SDP
Catalog No. 5000432547
Encompass_Preferred
Carvedilol phosphate hemihydrate is a non-selective β/α-1 blocker and a multiple action antihypertensive agent. It has potential applications in angina and congestive heart failure. This compound also acts as an autophagy inducer and inhibits the NLRP3 inflammasome.
  • Non-selective β/α-1 blocker
  • Inhibits lipid peroxidation with an IC50 of 5 μM
  • Multiple action antihypertensive agent
  • Potential applications in angina and congestive heart failure
  • Autophagy inducer
  • Inhibits NLRP3 inflammasome
  • Inhibits superoxide generation by activated human neutrophils in vitro with an IC50 of 28 μM
  • Scavenges oxygen free radicals in a cell-free system with an IC50 of 25 μM
Catalog No. 50-004-32547 Supplier Medchemexpress LLC Supplier No. HYB0006A1G
Carvedilol phosphate hemihydrate is a non-selective β/α-1 blocker and a multiple action antihypertensive agent. It has potential applications in angina and congestive heart failure. This compound also acts as an autophagy inducer and inhibits the NLRP3 inflammasome.
  • Non-selective β/α-1 blocker
  • Inhibits lipid peroxidation with an IC50 of 5 μM
  • Multiple action antihypertensive agent
  • Potential applications in angina and congestive heart failure
  • Autophagy inducer
  • Inhibits NLRP3 inflammasome
  • Inhibits superoxide generation by activated human neutrophils in vitro with an IC50 of 28 μM
  • Scavenges oxygen free radicals in a cell-free system with an IC50 of 25 μM