Learn More
Medchemexpress LLC Carvedilol phosphate hemihydrate | 610309-89-2 | 99.9% | C24H26N2O4.H3PO4.1/2H2O | 100 MG
SDP
Catalog No. 5000433346
Encompass_Preferred
Carvedilol phosphate hemihydrate is a non-selective β/α-1 blocker. It inhibits lipid peroxidation with an IC50 of 5 μM. It is a multiple action antihypertensive agent with potential use in angina and congestive heart failure. It is also an autophagy inducer that inhibits the NLRP3 inflammasome.
  • Non-selective β/α-1 blocker
  • Inhibits lipid peroxidation with an IC50 of 5 μM
  • Multiple action antihypertensive agent
  • Potential use in angina and congestive heart failure
  • Autophagy inducer
  • Inhibits the NLRP3 inflammasome
Catalog No. 50-004-33346 Supplier Medchemexpress LLC Supplier No. HYB0006A100MG
Carvedilol phosphate hemihydrate is a non-selective β/α-1 blocker. It inhibits lipid peroxidation with an IC50 of 5 μM. It is a multiple action antihypertensive agent with potential use in angina and congestive heart failure. It is also an autophagy inducer that inhibits the NLRP3 inflammasome.
  • Non-selective β/α-1 blocker
  • Inhibits lipid peroxidation with an IC50 of 5 μM
  • Multiple action antihypertensive agent
  • Potential use in angina and congestive heart failure
  • Autophagy inducer
  • Inhibits the NLRP3 inflammasome