Learn More
Medchemexpress LLC Carvedilol phosphate hemihydrate | 610309-89-2 | CB91870399 | 99.9% | C24H26N2O4.H3PO4.1/2H2O | 50 MG
SDP
Catalog No. 5000432937
Encompass_Preferred
Carvedilol phosphate hemihydrate (BM 14190 phosphate hemihydrate) is a non-selective β/α-1 blocker. It inhibits lipid peroxidation with an IC50 of 5 μM and acts as a multiple-action antihypertensive agent with potential uses in angina and congestive heart failure. Additionally, it is an autophagy inducer that inhibits the NLRP3 inflammasome.
  • Non-selective β/α-1 blocker.
  • Inhibits lipid peroxidation with an IC50 of 5 μM.
  • Multiple action antihypertensive agent.
  • Potential use in angina and congestive heart failure.
  • Autophagy inducer.
  • Inhibits the NLRP3 inflammasome.
Catalog No. 50-004-32937 Supplier Medchemexpress LLC Supplier No. HYB0006A50MG
Carvedilol phosphate hemihydrate (BM 14190 phosphate hemihydrate) is a non-selective β/α-1 blocker. It inhibits lipid peroxidation with an IC50 of 5 μM and acts as a multiple-action antihypertensive agent with potential uses in angina and congestive heart failure. Additionally, it is an autophagy inducer that inhibits the NLRP3 inflammasome.
  • Non-selective β/α-1 blocker.
  • Inhibits lipid peroxidation with an IC50 of 5 μM.
  • Multiple action antihypertensive agent.
  • Potential use in angina and congestive heart failure.
  • Autophagy inducer.
  • Inhibits the NLRP3 inflammasome.