Learn More
Medchemexpress LLC Carvedilol phosphate hemihydrate | 610309-89-2 | MFCD13194860 | 99.9% | C24H26N2O4.H3PO4.1/2H2O | 25 MG
SDP
Catalog No. 5000358886
Encompass_Preferred
Carvedilol phosphate hemihydrate is a non-selective β/α-1 blocker with multiple actions, including inhibiting lipid peroxidation with an IC50 of 5 μM. This compound acts as an antihypertensive agent with potential use in angina and congestive heart failure. Additionally, it functions as an autophagy inducer that inhibits the NLRP3 inflammasome.
  • Non-selective β/α-1 blocker
  • Inhibits lipid peroxidation
  • Antihypertensive agent
  • Potential use in angina and congestive heart failure
  • Autophagy inducer
  • Inhibits NLRP3 inflammasome
  • Inhibits superoxide generation by activated human neutrophils (IC50 of 28 μM)
  • Scavenges oxygen free radicals in a cell-free system (IC50 of 25 μM)
Catalog No. 50-003-58886 Supplier Medchemexpress LLC Supplier No. 25MGHYB0006A
Carvedilol phosphate hemihydrate is a non-selective β/α-1 blocker with multiple actions, including inhibiting lipid peroxidation with an IC50 of 5 μM. This compound acts as an antihypertensive agent with potential use in angina and congestive heart failure. Additionally, it functions as an autophagy inducer that inhibits the NLRP3 inflammasome.
  • Non-selective β/α-1 blocker
  • Inhibits lipid peroxidation
  • Antihypertensive agent
  • Potential use in angina and congestive heart failure
  • Autophagy inducer
  • Inhibits NLRP3 inflammasome
  • Inhibits superoxide generation by activated human neutrophils (IC50 of 28 μM)
  • Scavenges oxygen free radicals in a cell-free system (IC50 of 25 μM)