Learn More
Med Vet International Miltex Wax Carver, #3S, Each, 73-38
Catalog No. 502830533
Encompass_Preferred
Miltex wax carvers are used to carve and cut wax during impressions and various other dental applications which involve dental wax. Porcelain carvers are used for carving porcelain material during inlay, onlay and crown preparations. #3S.
Catalog No. 50-283-0533 Supplier Med Vet International Supplier No. MIL7338
Miltex wax carvers are used to carve and cut wax during impressions and various other dental applications which involve dental wax. Porcelain carvers are used for carving porcelain material during inlay, onlay and crown preparations. #3S.