Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (-)-Carvone | 6485-40-1 | 99.9% | 150.22 | C10H14O | 25 G
SDP

Catalog No. 5000304161
Encompass_Preferred

(-)-Carvone is the levorotatory enantiomer of carvone supplied as a high-purity research chemical. It is reported to act as an insect neurotoxin and an irreversible acetylcholinesterase inhibitor and is intended for laboratory research use only.

  • High purity (99.91%).
  • Molecular formula C10H14O; molecular weight 150.22.
  • Available as 10 mM solution in DMSO and as bulk solids including 25 g.
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • For research use only; not intended for human or veterinary applications.
  • Soluble in DMSO; provided ready-to-use as a 10 mM solution in some formats.

Catalog No. 50-003-04161 Supplier Medchemexpress LLC Supplier No. 25GHYW017423
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(-)-Carvone is the levorotatory enantiomer of carvone supplied as a high-purity research chemical. It is reported to act as an insect neurotoxin and an irreversible acetylcholinesterase inhibitor and is intended for laboratory research use only.

  • High purity (99.91%).
  • Molecular formula C10H14O; molecular weight 150.22.
  • Available as 10 mM solution in DMSO and as bulk solids including 25 g.
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • For research use only; not intended for human or veterinary applications.
  • Soluble in DMSO; provided ready-to-use as a 10 mM solution in some formats.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.