Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (-)-Carvone | 6485-40-1 | MFCD00001578 | 99.9% | 150.22 g/mol | C10H14O | 50 G
SDP

Catalog No. 5000301753
Encompass_Preferred

(-)-Carvone is the levorotatory enantiomer of carvone, a monoterpenoid found in spearmint and caraway essential oils. It is supplied as a high-purity research reagent suitable for in vitro and in vivo studies, with documented solubility protocols and storage recommendations; consult the SDS for safety and handling.

  • High purity: 99.91%.
  • Chemical formula C10H14O; molecular weight 150.22 g/mol.
  • Appearance: colorless to light yellow liquid; density ≈0.959 g/cm3.
  • Soluble in DMSO; in vitro and in vivo formulation protocols available.
  • Storage: recommended cold storage with defined shelf life in pure form and in solution.
  • Intended for research use only; safety data sheet available.

Catalog No. 50-003-01753 Supplier Medchemexpress LLC Supplier No. HYW01742350G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(-)-Carvone is the levorotatory enantiomer of carvone, a monoterpenoid found in spearmint and caraway essential oils. It is supplied as a high-purity research reagent suitable for in vitro and in vivo studies, with documented solubility protocols and storage recommendations; consult the SDS for safety and handling.

  • High purity: 99.91%.
  • Chemical formula C10H14O; molecular weight 150.22 g/mol.
  • Appearance: colorless to light yellow liquid; density ≈0.959 g/cm3.
  • Soluble in DMSO; in vitro and in vivo formulation protocols available.
  • Storage: recommended cold storage with defined shelf life in pure form and in solution.
  • Intended for research use only; safety data sheet available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.