Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich ™-(-)-Carvone

Catalog No. 124931500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-493-1500ML 500 mL
12-493-1100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 12-493-1500ML Supplier Sigma Aldrich Supplier No. 124931500ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(R)-(-)-Carvone is a monoterpenoid. Its ozonolysis in the gaseous phase has been proposed. Aldehydes (formaldehyde) and biradicals were obtained as the major products. It participates in the diastereoselective synthesis of homochiral octalones.Chiral starting material.

Specifications

CAS 6485-40-1
Density 0.959 g/mL at 25°C
Boiling Point 227°C to 230°C
Molecular Formula C10H14O
Refractive Index n20/D 1.497
Linear Formula C10H14O
MDL Number MFCD00001578
Quantity 500 mL
Synonym (-)-Carvone; (R)-5-Isopropenyl-2-methyl-2-cyclohexenone; p-Mentha-6,8-dien-2-one; Carvol
Molecular Weight (g/mol) 150.22 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.