Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-Carvone, ≥98.5% (sum of enantiomers, GC), MilliporeSigma™ Supelco™

Catalog No. 111013586 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
25 mL
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3586 1 mL
11-101-3584 25 mL
11-101-3585 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-3586 Supplier MilliporeSigma Supelco Supplier No. 220701ML
Add to Cart
Add to Cart
Featured Industries: Agriculture and Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

CAS 2244-16-8
Density 0.960 g/mL (at 20°C)
Flash Point 89°C - Closed Cup
Physical Form Neat
Linear Formula C10H14 O
MDL Number MFCD00062997
Boiling Point 228°C to 230°C
Synonym (+)-p-Mentha-6,8-diene 2-one; (S)-5-Isopropenyl-2-methyl-2-cyclohexenone
Type Analytical Standard
Quantity 1 mL
Formula Weight 150.22
Percent Purity ≥98.5% (sum of enantiomers, GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.