Learn More
Cayman Chemical Caryophylen oxIde 1g
SDP
Catalog No. 502986385
Encompass_Preferred
A bicyclic sesquiterpene; a metabolite of β-caryophyllene that has been found in C. sativa; does not bind to the cannabinoid (CB) receptor CB2 (Ki = >20 µM); induces calcium currents in HL-60 cells expressing CB2 receptors but also does so in CB2-deficient cells; cytotoxic to HepG2, AGS, HeLa, SNU-1 and SNU-16 cells (IC50s = 3.95, 12.6, 13.55, 16.79, and 27.39 µM, respectively); the volatile component of C. sativa sensed by narcotic detection dogs
Catalog No. 50-298-6385 Supplier Cayman Chemical Supplier No. 215731 G
A bicyclic sesquiterpene; a metabolite of β-caryophyllene that has been found in C. sativa; does not bind to the cannabinoid (CB) receptor CB2 (Ki = >20 µM); induces calcium currents in HL-60 cells expressing CB2 receptors but also does so in CB2-deficient cells; cytotoxic to HepG2, AGS, HeLa, SNU-1 and SNU-16 cells (IC50s = 3.95, 12.6, 13.55, 16.79, and 27.39 µM, respectively); the volatile component of C. sativa sensed by narcotic detection dogs