Learn More
Cayman Chemical β-Caryophyllene, Purity: ≥95%, CAS: 87-44-5, 100 mg
SDP
Catalog No. 502987435
Encompass_Preferred
A sesquiterpene with diverse biological activities; an agonist of the CB2 receptor (Ki = 155 nM) that inhibits forskolin-induced cAMP production in CHO-K1 cells expressing CB2 receptors (EC50 = 38 nM); an agonist of PPARα (EC50 = 9.58 µM in a reporter assay); reduces MPP+-induced production of ROS in and protects against cytotoxicity of SH-SY5Y cells at 1 and 2.5 µM; decreases the β-amyloid burden in the hippocampus and cerebral cortex and improves memory in an APP/PS1 transgenic mouse model of Alzheimer’s disease at 48 mg/kg; has anxiolytic-like and antidepressant-like activity in mice at 50 mg/kg
Catalog No. 50-298-7435 Supplier Cayman Chemical Supplier No. 21572100 MG
A sesquiterpene with diverse biological activities; an agonist of the CB2 receptor (Ki = 155 nM) that inhibits forskolin-induced cAMP production in CHO-K1 cells expressing CB2 receptors (EC50 = 38 nM); an agonist of PPARα (EC50 = 9.58 µM in a reporter assay); reduces MPP+-induced production of ROS in and protects against cytotoxicity of SH-SY5Y cells at 1 and 2.5 µM; decreases the β-amyloid burden in the hippocampus and cerebral cortex and improves memory in an APP/PS1 transgenic mouse model of Alzheimer’s disease at 48 mg/kg; has anxiolytic-like and antidepressant-like activity in mice at 50 mg/kg