Learn More
Medchemexpress LLC HY-N1415 500mg Medchemexpress, β-Caryophyllene CAS:87-44-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN1415500MG
Encompass_Preferred
Medchemexpress, HY-N1415 500mg β-Caryophyllene CAS:87-44-5 β-Caryophyllene is a CB2 receptor agonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3825