Learn More
Selleck Chemical LLC Caryophyllene oxide S3983-25mg
SDP
Supplier:  Selleck Chemical LLC S398325MG
Encompass_Preferred
Caryophyllene oxide an oxygenated terpenoid existed in many plant essential oil is well known as preservative in food drugs and cosmetics with anti-inflammatory anti-carcinogenic and skin penetration enhancing properties
Catalog No. 50-310-9468