Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-Caryophyllene oxide, ≥99% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012639 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2639 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2639 Supplier MilliporeSigma Supelco Supplier No. 91034100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Analytical standard used in the quality control of flavor and fragrance components in food and cosmetic products.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Caryophyllene Oxide
CAS 1139-30-6
Flash Point 110°C
Physical Form Neat
Linear Formula C15H24 O
Technique Gas Chromatography
MDL Number MFCD00134216
Melting Point 62°C to 63°C
Synonym beta-Caryophyllene epoxide; (-)-Epoxycaryophyllene; (-)-Epoxydihydrocaryophyllene; (1 R,4 R,6 R,10 S)-9-Methylene-4,12,12-trimethyl-5-oxatricyclo[8.2.0.04,6 ]dodecane; trans-Caryophyllene oxide
Type Analytical Standard
Molecular Weight (g/mol) 220.35
Industry Type Flavors and Fragrances
Quantity 100 mg
Formula Weight 220.35
Percent Purity ≥99% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.