Learn More
System Biosciences LLC CAS 9 RT PCR PRIMER SET50 µL at 5 µM for each primer set
SDP
Catalog No. NC1129850
Encompass_Preferred
With our ready-to-go Cas9 RT-PCR Primer Set, you can quickly and easily confirm Cas9 expression in cells or in vitro transcription assays.The amplicon for Primer Set 1 is 219bp and amplicon for Primer Set 2 is 122bp. These primer sets allow for detection of Cas9 nuclease, Cas9 Nickase and Cas9 double mutant at messenger level. The primer sets are compatible with Cas9 mRNA expression from any SBI Cas9 SmartNuclease construct and vectors
Catalog No. NC1129850 Supplier System Biosciences LLC Supplier No. CAS9PR1
With our ready-to-go Cas9 RT-PCR Primer Set, you can quickly and easily confirm Cas9 expression in cells or in vitro transcription assays.The amplicon for Primer Set 1 is 219bp and amplicon for Primer Set 2 is 122bp. These primer sets allow for detection of Cas9 nuclease, Cas9 Nickase and Cas9 double mutant at messenger level. The primer sets are compatible with Cas9 mRNA expression from any SBI Cas9 SmartNuclease construct and vectors