Learn More
INTACT GENOMICS INC Cas12a Nuclease 40 µg
Catalog No. 5000082961
Encompass
ApplicationsIntact Genomics (ig) Cas12a Nuclease is the purified recombinant Francisella tularensis Cas12a enzyme for in vitro editing. This enzyme is designed to perform CRISPR/Cas12a-mediated genome editing. The physical purity of this enzyme is ≥65% as assessed by SDS-PAGE with Coomassie blue staining and densitometry.Product Components: Cas12a Nuclease 10x Cas12a Nuclease Reaction Buffer
Catalog No. 50-000-82961 Supplier INTACT GENOMICS INC Supplier No. 3370
ApplicationsIntact Genomics (ig) Cas12a Nuclease is the purified recombinant Francisella tularensis Cas12a enzyme for in vitro editing. This enzyme is designed to perform CRISPR/Cas12a-mediated genome editing. The physical purity of this enzyme is ≥65% as assessed by SDS-PAGE with Coomassie blue staining and densitometry.Product Components: Cas12a Nuclease 10x Cas12a Nuclease Reaction Buffer