Learn More
INTACT GENOMICS INC Cas12a Nuclease 400 µg
Catalog No. 5000082873
Encompass
Intact Genomics (ig) Cas12a Nuclease is the purified recombinant Francisella tularensis Cas12a enzyme for in vitro editing. This enzyme is designed to perform CRISPR/Cas12a-mediated genome editing. The physical purity of this enzyme is ≥65% as assessed by SDS-PAGE with Coomassie blue staining and densitometry.Product Components: Cas12a Nuclease 10x Cas12a Nuclease Reaction Buffer
Catalog No. 50-000-82873 Supplier INTACT GENOMICS INC Supplier No. 3376
Intact Genomics (ig) Cas12a Nuclease is the purified recombinant Francisella tularensis Cas12a enzyme for in vitro editing. This enzyme is designed to perform CRISPR/Cas12a-mediated genome editing. The physical purity of this enzyme is ≥65% as assessed by SDS-PAGE with Coomassie blue staining and densitometry.Product Components: Cas12a Nuclease 10x Cas12a Nuclease Reaction Buffer