Learn More
Sigma Aldrich Fine Chemicals Biosciences CAS9 Blasticidin Lenti Plasmid, 50 uL vial
Catalog No. NC1813139
Encompass
This product is a blasticidin resistant lenti-Cas9 plasmid for generation of lentiviral particles. Functional Genomics/Screening /Target Validation/Genome Editing.
Catalog No. NC1813139 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CAS9BST1EA
This product is a blasticidin resistant lenti-Cas9 plasmid for generation of lentiviral particles. Functional Genomics/Screening /Target Validation/Genome Editing.