Learn More
Sigma Aldrich Fine Chemicals Biosciences Cas9 D10A Nickase Protein
Catalog No. 502906011
Encompass
Recombinant Cas9-D10A Nickase protein from Streptococcus pyogenes (~160 KD) is a ready-to-use reagent for genome engineering experiments. When combined with target-specific paired guide RNAs S. pyogenes Cas9-D10A nickase will act as a targeted nuclease suitable for transfection of cell cultures and for the accelerated development of genetically-modified animals via one-cell embryo injection.
Catalog No. 50-290-6011 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CAS9D10APR250UG
Recombinant Cas9-D10A Nickase protein from Streptococcus pyogenes (~160 KD) is a ready-to-use reagent for genome engineering experiments. When combined with target-specific paired guide RNAs S. pyogenes Cas9-D10A nickase will act as a targeted nuclease suitable for transfection of cell cultures and for the accelerated development of genetically-modified animals via one-cell embryo injection.