Learn More
XpressBio Cas9 ELISA Assay, 96 wells
SDP
Supplier:  XpressBio CAS91000
Encompass_Preferred
The assay can be used to monitor S. pyogenes Cas9 in cell culture lysate and is ideal for screening experimental samples utilizing the CRISPR/Cas9 gene editing system.
Catalog No. NC1722909