Learn More
Sigma Aldrich Fine Chemicals Biosciences Cas9 GFP Lentiviral Partic
Catalog No. 502907096
Encompass
Ready-to-use Cas9-GFP lentiviral particles enable immediate transduction of a wide range of cell lines. Cas9-GFP lentiviral particles efficiently and stably integrate Cas9 and GFP linked by a 2A peptide and driven by the EF1 alpha promoter (EF1a-Cas9-2A-GFP) for strong expression in both dividing and non-dividing cell lines. They are ideal for avoiding the tedious lentivirus production process and are one part of a two part CRISPR system with individual Cas9 and gRNA expression vectors. For easy fluorescence microscopy a MOI greater than 200 should be employed. To order gRNA in any format click here
Catalog No. 50-290-7096 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CAS9GFPV200UL
Ready-to-use Cas9-GFP lentiviral particles enable immediate transduction of a wide range of cell lines. Cas9-GFP lentiviral particles efficiently and stably integrate Cas9 and GFP linked by a 2A peptide and driven by the EF1 alpha promoter (EF1a-Cas9-2A-GFP) for strong expression in both dividing and non-dividing cell lines. They are ideal for avoiding the tedious lentivirus production process and are one part of a two part CRISPR system with individual Cas9 and gRNA expression vectors. For easy fluorescence microscopy a MOI greater than 200 should be employed. To order gRNA in any format click here