Learn More
Enzo Life Sciences Cas9 monoclonal antibody (7A9) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS2940100
Encompass_Preferred
Clone: 7A9. Isotype: IgG1κ. Immunogen: N-terminal fragment of Streptococcus pyogenes Cas9. Source: Purified from mouse ascites. Specificity: Recognizes both Cas9 and dCas9 (nuclease deficient Cas9). Applications: IF, IP, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-201-2834