Learn More
OriGene CAS9 MOUSE MONOCLONAL 100UL
SDP
Supplier:  OriGene TA811179AM
Encompass_Preferred
CAS9 mouse monoclonal antibody,clone OTI5E5, Biotinylated Immunogen Human recombinant protein fragment corresponding to amino acids 1-1166 of human CAS9 produced in E.coli.
Catalog No. NC2603067