Learn More
Apexbio Technology LLC SpCas9 mRNA (ARCA 5mCTP ψUTP) 1mg (1mg/mL)
SDP
Catalog No. 503107164
Encompass_Preferred
SpCas9 mRNA (ARCA 5mCTP UTP) encodes Streptococcus pyogenes Cas9 (SpCas9) protein This product can be used with purified guide RNA (gRNA) sequences in the CRISPR/Cas system for recognizing and cleaving genomic DNA targets CRISPR/Cas a defense system has been adapted from the Type II CRISPR/Cas system of S pyogenes for eukaryotic gene editing It requires two key components the Cas9 endonuclease and a non-coding gRNA Guided by a single gRNA Cas9 introduces DNA double-strand breaks (DSBs) at specific genomic sites Cells activate endogenous DNA repair pathways non-homologous end joining (NHEJ) or homology-directed repair (HDR) to repair the targeted DSB SpCas9 mRNA (ARCA 5mCTP UTP)is designed to mimick mature eukaryotic mRNA It contains a cap 0 structure generated via co-transcriptional capping Incorporation of modified nucleotides 5mCTP UTP poly (A) tail minimizes RNA-induced innate immune activation and enhances mRNA stability and lifespan both in vitro and in vivo
Catalog No. 50-310-7164 Supplier Apexbio Technology LLC Supplier No. R10061.0
SpCas9 mRNA (ARCA 5mCTP UTP) encodes Streptococcus pyogenes Cas9 (SpCas9) protein This product can be used with purified guide RNA (gRNA) sequences in the CRISPR/Cas system for recognizing and cleaving genomic DNA targets CRISPR/Cas a defense system has been adapted from the Type II CRISPR/Cas system of S pyogenes for eukaryotic gene editing It requires two key components the Cas9 endonuclease and a non-coding gRNA Guided by a single gRNA Cas9 introduces DNA double-strand breaks (DSBs) at specific genomic sites Cells activate endogenous DNA repair pathways non-homologous end joining (NHEJ) or homology-directed repair (HDR) to repair the targeted DSB SpCas9 mRNA (ARCA 5mCTP UTP)is designed to mimick mature eukaryotic mRNA It contains a cap 0 structure generated via co-transcriptional capping Incorporation of modified nucleotides 5mCTP UTP poly (A) tail minimizes RNA-induced innate immune activation and enhances mRNA stability and lifespan both in vitro and in vivo